Assay Details
Amplicon Length: | 91 |
Chromosome Location: | Chr. 1 - 228397266 - 228397266 [+] on 228397266 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6742-3p | MIMAT0027386 |
Human | hsa-miR-6742-5p | MIMAT0027385 |
Stem-loop Details
Human