Assay Details
Mature miRNA Sequence: | AGUGGGGUGGGACCCAGCUGUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6742-5p | MIMAT0027385 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-6742-5p | ||
---|---|---|---|
miRBase Accession Number: | MI0022587 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.1: 228397048 - 228397109 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | AGUGGGGUGGGACCCAGCUGUU | ||
Species: | Human | ||
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Mature miRNA Sequence: | AGUGGGGUGGGACCCAGCUGUU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6742-5p | MIMAT0027385 |
Stem-loop ID | hsa-mir-6742 |
Stem-loop Accession # | MI0022587 |
Stem-loop Sequence | GAGGGAGUGGGGUGGGACCCAGCUGUUGGCCAUGGCGACAACACCUGGGUUGUCCCCUCUAG |
Chromosome Location | Chr. 1 - 228397048 - 228397109 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0027385
miRBase ID: hsa-miR-6742-5p Mature miRNA Sequence: AGUGGGGUGGGACCCAGCUGUU Chromosome Location: Chr. 1 - 228397048 - 228397109 [-] on Build GRCh38 |