Assay Details
Amplicon Length: | 87 |
Chromosome Location: | Chr. 19 - 6392736 - 6392736 [+] on 6392736 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6790-3p | MIMAT0027481 |
Human | hsa-miR-6790-5p | MIMAT0027480 |
Stem-loop Details
Human