Assay Details
Gene Family ID: | MIPF0001811, mir-6790 |
Mature miRNA Sequence: | CGACCUCGGCGACCCCUCACU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6790-3p | MIMAT0027481 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-6790-3p | ||
---|---|---|---|
miRBase Accession Number: | MI0022635 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.19: 6392921 - 6392983 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | CGACCUCGGCGACCCCUCACU | ||
Species: | Human | ||
Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0001811, mir-6790 |
Mature miRNA Sequence: | CGACCUCGGCGACCCCUCACU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6790-3p | MIMAT0027481 |
Stem-loop ID | hsa-mir-6790 |
Stem-loop Accession # | MI0022635 |
Stem-loop Sequence | GUGAGUGUGGAUUUGGCGGGGUUCGGGGGUUCCGACGGCGACCUCGGCGACCCCUCACUCACC |
Chromosome Location | Chr. 19 - 6392921 - 6392983 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0027481
miRBase ID: hsa-miR-6790-3p Mature miRNA Sequence: CGACCUCGGCGACCCCUCACU Chromosome Location: Chr. 19 - 6392921 - 6392983 [-] on Build GRCh38 |