Assay Details
Amplicon Length: | 74 |
Chromosome Location: | Chr. 10 - 641511 - 641511 [+] on 641511 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-5699-3p | MIMAT0022492 |
Human | hsa-miR-5699-5p | MIMAT0027103 |
Stem-loop Details
Human