Assay Details
Mature miRNA Sequence: | UGCCCCAACAAGGAAGGACAAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-5699-5p | MIMAT0027103 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-5699-5p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0019306 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.10: 641689 - 641778 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | UGCCCCAACAAGGAAGGACAAG | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | UGCCCCAACAAGGAAGGACAAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-5699-5p | MIMAT0027103 |
Stem-loop ID | hsa-mir-5699 |
Stem-loop Accession # | MI0019306 |
Stem-loop Sequence | CUGUACCCCUGCCCCAACAAGGAAGGACAAGAGGUGUGAGCCACACACACGCCUGGCCUCCUGUCUUUCCUUGUUGGAGCAGGGAUGUAG |
Chromosome Location | Chr. 10 - 641689 - 641778 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0027103
miRBase ID: hsa-miR-5699-5p Mature miRNA Sequence: UGCCCCAACAAGGAAGGACAAG Chromosome Location: Chr. 10 - 641689 - 641778 [-] on Build GRCh38 |