Assay Details
Amplicon Length: | 64 |
Chromosome Location: | Chr. 9 - 20410995 - 20410995 [-] on 20410995 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4473 | MIMAT0019000 |
Stem-loop Details
Human