Assay Details
Mature miRNA Sequence: | CUAGUGCUCUCCGUUACAAGUA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4473 | MIMAT0019000 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4473 | ||
---|---|---|---|
miRBase Accession Number: | MI0016825 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.9: 20411148 - 20411238 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | CUAGUGCUCUCCGUUACAAGUA | ||
Species: | Human | ||
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Mature miRNA Sequence: | CUAGUGCUCUCCGUUACAAGUA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4473 | MIMAT0019000 |
Stem-loop ID | hsa-mir-4473 |
Stem-loop Accession # | MI0016825 |
Stem-loop Sequence | AAGGAACAGGGGACACUUGUAAUGGAGAACACUAAGCUAUGGACUGCUAUGGACUGCUAGUGCUCUCCGUUACAAGUAUCCCCUGUUACCU |
Chromosome Location | Chr. 9 - 20411148 - 20411238 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019000
miRBase ID: hsa-miR-4473 Mature miRNA Sequence: CUAGUGCUCUCCGUUACAAGUA Chromosome Location: Chr. 9 - 20411148 - 20411238 [-] on Build GRCh38 |