Assay Details
Amplicon Length: | 72 |
Chromosome Location: | Chr. 8 - 95072564 - 95072564 [+] on 95072564 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3150a-5p | MIMAT0019206 |
Human | hsa-miR-3150a-3p | MIMAT0015023 |
Human | hsa-miR-3150b-5p | MIMAT0019226 |
Human | hsa-miR-3150b-3p | MIMAT0018194 |
Stem-loop Details
Human