Assay Details
Gene Family ID: | MIPF0001102, mir-3150 |
Mature miRNA Sequence: | CUGGGGAGAUCCUCGAGGUUGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3150a-3p | MIMAT0015023 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-3150a-3p | ||
---|---|---|---|
miRBase Accession Number: | MI0014177 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.8: 95072914 - 95072993 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | CUGGGGAGAUCCUCGAGGUUGG | ||
Species: | Human | ||
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0001102, mir-3150 |
Mature miRNA Sequence: | CUGGGGAGAUCCUCGAGGUUGG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3150a-3p | MIMAT0015023 |
Stem-loop ID | hsa-mir-3150a |
Stem-loop Accession # | MI0014177 |
Stem-loop Sequence | GGGAAGCAGGCCAACCUCGACGAUCUCCUCAGCACCUGAACGCCAAGGCUGGGGAGAUCCUCGAGGUUGGCCUGCUUUCC |
Chromosome Location | Chr. 8 - 95072914 - 95072993 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0015023
miRBase ID: hsa-miR-3150a-3p Mature miRNA Sequence: CUGGGGAGAUCCUCGAGGUUGG Chromosome Location: Chr. 8 - 95072914 - 95072993 [+] on Build GRCh38 |