Assay Details
Amplicon Length: | 94 |
Chromosome Location: | Chr. 22 - 48274206 - 48274206 [+] on 48274206 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3201 | MIMAT0015086 |
Stem-loop Details
Human