Assay Details
Mature miRNA Sequence: | GGGAUAUGAAGAAAAAU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3201 | MIMAT0015086 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-3201 | ||
---|---|---|---|
Stem-loop Accession Number: | MI0014250 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.22: 48274364 - 48274415 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | GGGAUAUGAAGAAAAAU | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | GGGAUAUGAAGAAAAAU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3201 | MIMAT0015086 |
Stem-loop ID | hsa-mir-3201 |
Stem-loop Accession # | MI0014250 |
Stem-loop Sequence | GGGAUAUGAAGAAAAAUAAGAGGCUAGGAUUGCCUCUUAUUUUUACAUGCCC |
Chromosome Location | Chr. 22 - 48274364 - 48274415 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0015086
miRBase ID: hsa-miR-3201 Mature miRNA Sequence: GGGAUAUGAAGAAAAAU Chromosome Location: Chr. 22 - 48274364 - 48274415 [+] on Build GRCh38 |