Assay Details
Amplicon Length: | 81 |
Chromosome Location: | Chr. 15 - 59171038 - 59171038 [+] on 59171038 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-2116-3p | MIMAT0011161 |
Human | hsa-miR-2116-5p | MIMAT0011160 |
Stem-loop Details
Human