Assay Details
Mature miRNA Sequence: | CCUCCCAUGCCAAGAACUCCC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-2116-3p | MIMAT0011161 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-2116-3p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0010635 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.15: 59171183 - 59171262 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | CCUCCCAUGCCAAGAACUCCC | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | CCUCCCAUGCCAAGAACUCCC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-2116-3p | MIMAT0011161 |
Stem-loop ID | hsa-mir-2116 |
Stem-loop Accession # | MI0010635 |
Stem-loop Sequence | GACCUAGGCUAGGGGUUCUUAGCAUAGGAGGUCUUCCCAUGCUAAGAAGUCCUCCCAUGCCAAGAACUCCCAGACUAGGA |
Chromosome Location | Chr. 15 - 59171183 - 59171262 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0011161
miRBase ID: hsa-miR-2116-3p Mature miRNA Sequence: CCUCCCAUGCCAAGAACUCCC Chromosome Location: Chr. 15 - 59171183 - 59171262 [-] on Build GRCh38 |