Assay Details
Gene Family ID: | MIPF0001578, mir-4774 |
Mature miRNA Sequence: | AUUGCCUAACAUGUGCCAGAA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4774-3p | MIMAT0019930 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4774-3p | ||
---|---|---|---|
miRBase Accession Number: | MI0017417 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.2: 168582943 - 168583018 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | AUUGCCUAACAUGUGCCAGAA | ||
Species: | Human | ||
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0001578, mir-4774 |
Mature miRNA Sequence: | AUUGCCUAACAUGUGCCAGAA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4774-3p | MIMAT0019930 |
Stem-loop ID | hsa-mir-4774 |
Stem-loop Accession # | MI0017417 |
Stem-loop Sequence | UAUAUUGUUGUCUGGUAUGUAGUAGGUAAUAACUGACAAACAGACAAUUGCCUAACAUGUGCCAGAAAACAACAUA |
Chromosome Location | Chr. 2 - 168582943 - 168583018 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019930
miRBase ID: hsa-miR-4774-3p Mature miRNA Sequence: AUUGCCUAACAUGUGCCAGAA Chromosome Location: Chr. 2 - 168582943 - 168583018 [+] on Build GRCh38 |