Assay Details
Amplicon Length: | 97 |
Chromosome Location: | Chr. 2 - 168582792 - 168582792 [+] on 168582792 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4774-3p | MIMAT0019930 |
Human | hsa-miR-4774-5p | MIMAT0019929 |
Stem-loop Details
Human