Assay Details
Mature miRNA Sequence: | UGGGGGAGUGCAGUGAUUGUGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-5698 | MIMAT0022491 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-5698 | ||
---|---|---|---|
miRBase Accession Number: | MI0019305 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.1: 154104521 - 154104592 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | UGGGGGAGUGCAGUGAUUGUGG | ||
Species: | Human | ||
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Mature miRNA Sequence: | UGGGGGAGUGCAGUGAUUGUGG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-5698 | MIMAT0022491 |
Stem-loop ID | hsa-mir-5698 |
Stem-loop Accession # | MI0019305 |
Stem-loop Sequence | CUGUGCACCUGGGGGAGUGCAGUGAUUGUGGAAUGCAAAGUCCCACAAUCACUGUACUCCCCAGGUGCACAG |
Chromosome Location | Chr. 1 - 154104521 - 154104592 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0022491
miRBase ID: hsa-miR-5698 Mature miRNA Sequence: UGGGGGAGUGCAGUGAUUGUGG Chromosome Location: Chr. 1 - 154104521 - 154104592 [-] on Build GRCh38 |