Assay Details
Amplicon Length: | 95 |
Chromosome Location: | Chr. 1 - 154104231 - 154104231 [-] on 154104231 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-5698 | MIMAT0022491 |
Stem-loop Details
Human