Assay Details
Mature miRNA Sequence: | GCUGAACUGGGCUGAGCUGGGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4539 | MIMAT0019082 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4539 | ||
---|---|---|---|
miRBase Accession Number: | MI0016910 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.14: 105857513 - 105857572 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | GCUGAACUGGGCUGAGCUGGGC | ||
Species: | Human | ||
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Mature miRNA Sequence: | GCUGAACUGGGCUGAGCUGGGC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4539 | MIMAT0019082 |
Stem-loop ID | hsa-mir-4539 |
Stem-loop Accession # | MI0016910 |
Stem-loop Sequence | UGAGCUGAGCUCUGCUGUGCUGUGCUGAGCAGGGCUGAGCUGAACUGGGCUGAGCUGGGC |
Chromosome Location | Chr. 14 - 105857513 - 105857572 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019082
miRBase ID: hsa-miR-4539 Mature miRNA Sequence: GCUGAACUGGGCUGAGCUGGGC Chromosome Location: Chr. 14 - 105857513 - 105857572 [-] on Build GRCh38 |