Assay Details
Amplicon Length: | 103 |
Chromosome Location: | Chr. 14 - 105858674 - 105858674 [+] on 105858674 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4507 | MIMAT0019044 |
Human | hsa-miR-4537 | MIMAT0019080 |
Human | hsa-miR-4538 | MIMAT0019081 |
Human | hsa-miR-4539 | MIMAT0019082 |
Stem-loop Details
Human