Assay Details
Mature miRNA Sequence: | UAAUGCAUUAAAUUAUUGAAGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-5700 | MIMAT0022493 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-5700 | ||
---|---|---|---|
miRBase Accession Number: | MI0019307 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.12: 94561789 - 94561859 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | UAAUGCAUUAAAUUAUUGAAGG | ||
Species: | Human | ||
Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | UAAUGCAUUAAAUUAUUGAAGG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-5700 | MIMAT0022493 |
Stem-loop ID | hsa-mir-5700 |
Stem-loop Accession # | MI0019307 |
Stem-loop Sequence | UUAAUUAAUGCAUUAAAUUAUUGAAGGCCCUUGGGCACCCCAGGCCUUCAAUAAUUUAAUGCAUUUAUUGA |
Chromosome Location | Chr. 12 - 94561789 - 94561859 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0022493
miRBase ID: hsa-miR-5700 Mature miRNA Sequence: UAAUGCAUUAAAUUAUUGAAGG Chromosome Location: Chr. 12 - 94561789 - 94561859 [+] on Build GRCh38 |