Assay Details
Amplicon Length: | 99 |
Chromosome Location: | Chr. 12 - 94562110 - 94562110 [+] on 94562110 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-5700 | MIMAT0022493 |
Stem-loop Details
Human