Assay Details
Mature miRNA Sequence: | AUUUGUGCUUGGCUCUGUCAC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-2113 | MIMAT0009206 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-2113 | ||
---|---|---|---|
miRBase Accession Number: | MI0003939 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.6: 98024531 - 98024621 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | AUUUGUGCUUGGCUCUGUCAC | ||
Species: | Human | ||
Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | AUUUGUGCUUGGCUCUGUCAC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-2113 | MIMAT0009206 |
Stem-loop ID | hsa-mir-2113 |
Stem-loop Accession # | MI0003939 |
Stem-loop Sequence | UUUUCAAAGCAAUGUGUGACAGGUACAGGGACAAAUCCCGUUAAUAAGUAAGAGGAUUUGUGCUUGGCUCUGUCACAUGCCACUUUGAAAA |
Chromosome Location | Chr. 6 - 98024531 - 98024621 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0009206
miRBase ID: hsa-miR-2113 Mature miRNA Sequence: AUUUGUGCUUGGCUCUGUCAC Chromosome Location: Chr. 6 - 98024531 - 98024621 [+] on Build GRCh38 |