Assay Details
Amplicon Length: | 109 |
Chromosome Location: | Chr. 6 - 98024781 - 98024781 [-] on 98024781 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-2113 | MIMAT0009206 |
Stem-loop Details
Human