Assay Details
Amplicon Length: | 101 |
Chromosome Location: | Chr. 8 - 144792150 - 144792150 [-] on 144792150 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6850-3p | MIMAT0027601 |
Human | hsa-miR-6850-5p | MIMAT0027600 |
Stem-loop Details
Human