Assay Details
Mature miRNA Sequence: | GUGCGGAACGCUGGCCGGGGCG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6850-5p | MIMAT0027600 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-6850-5p | ||
---|---|---|---|
miRBase Accession Number: | MI0022696 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.8: 144791931 - 144791991 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | GUGCGGAACGCUGGCCGGGGCG | ||
Species: | Human | ||
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Mature miRNA Sequence: | GUGCGGAACGCUGGCCGGGGCG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6850-5p | MIMAT0027600 |
Stem-loop ID | hsa-mir-6850 |
Stem-loop Accession # | MI0022696 |
Stem-loop Sequence | GUGCGGAACGCUGGCCGGGGCGGGAGGGGAAGGGACGCCCGGCCGGAACGCCGCACUCACG |
Chromosome Location | Chr. 8 - 144791931 - 144791991 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0027600
miRBase ID: hsa-miR-6850-5p Mature miRNA Sequence: GUGCGGAACGCUGGCCGGGGCG Chromosome Location: Chr. 8 - 144791931 - 144791991 [-] on Build GRCh38 |