Assay Details
Amplicon Length: | 99 |
Chromosome Location: | Chr. 4 - 60922866 - 60922866 [-] on 60922866 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548ag | MIMAT0018969 |
Stem-loop Details
Human