Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | AAAGGUAAUUGUGGUUUCUGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548ag | MIMAT0018969 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-548ag | ||
---|---|---|---|
miRBase Accession Number: | MI0016793 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.4: 60922619 - 60922684 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | AAAGGUAAUUGUGGUUUCUGC | ||
Species: | Human | ||
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | AAAGGUAAUUGUGGUUUCUGC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548ag | MIMAT0018969 |
Stem-loop ID | hsa-mir-548ag-1 |
Stem-loop Accession # | MI0016793 |
Stem-loop Sequence | GUGCAAAGGUAAUUGUGGUUUCUGCUUUUAAAGGUAAUGGCAAAUAUUACAUUUACUUUUGCACCA |
Chromosome Location | Chr. 4 - 60922619 - 60922684 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0018969
miRBase ID: hsa-miR-548ag Mature miRNA Sequence: AAAGGUAAUUGUGGUUUCUGC Chromosome Location: Chr. 4 - 60922619 - 60922684 [+] on Build GRCh38 |
Stem-loop ID | hsa-mir-548ag-2 |
Stem-loop Accession # | MI0016794 |
Stem-loop Sequence | UGCAAAGGUAAUUGUGGUUUCUGCCAUUGAAAGUAAAGGCAAGAACCUCAAUUACCUUUGCAGC |
Chromosome Location | Chr. 20 - 60564562 - 60564625 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0018969
miRBase ID: hsa-miR-548ag Mature miRNA Sequence: AAAGGUAAUUGUGGUUUCUGC Chromosome Location: Chr. 4 - 60922619 - 60922684 [+] on Build GRCh38 |