Assay Details
Amplicon Length: | 108 |
Chromosome Location: | Chr. 12 - 48772305 - 48772305 [-] on 48772305 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4701-3p | MIMAT0019799 |
Human | hsa-miR-4701-5p | MIMAT0019798 |
Stem-loop Details
Human