Assay Details
Mature miRNA Sequence: | UUGGCCACCACACCUACCCCUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4701-5p | MIMAT0019798 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4701-5p | ||
---|---|---|---|
miRBase Accession Number: | MI0017334 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.12: 48771975 - 48772037 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | UUGGCCACCACACCUACCCCUU | ||
Species: | Human | ||
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Mature miRNA Sequence: | UUGGCCACCACACCUACCCCUU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4701-5p | MIMAT0019798 |
Stem-loop ID | hsa-mir-4701 |
Stem-loop Accession # | MI0017334 |
Stem-loop Sequence | CCUUGGCCACCACACCUACCCCUUGUGAAUGUCGGGCAAUGGGUGAUGGGUGUGGUGUCCACA |
Chromosome Location | Chr. 12 - 48771975 - 48772037 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019798
miRBase ID: hsa-miR-4701-5p Mature miRNA Sequence: UUGGCCACCACACCUACCCCUU Chromosome Location: Chr. 12 - 48771975 - 48772037 [-] on Build GRCh38 |