Invitrogen
Manufacturer Code:PA5102730
Catalog # PIPA5102730
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PTGES3 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5730442A20Rik Co chaperone p23 cPGES cytosolic prostaglandin E synthase cytosolic prostaglandin E2 synthase Hsp90 co chaperone Hsp90 co-chaperone LOC100345490 LOC100351480 P23 p23 cochaperone progesterone receptor complex p23 Prostaglandin E synthase 3 prostaglandin E synthase 3 (cytosolic) prostaglandin-E synthase 3 prostaglandin-E synthase 3-like Ptges PTGES3 ptges3 {ECO:0000250 RGD1561913 Sid 3177 Sid3177 Tebp telomerase binding protein p23 telomerase-binding protein p23 unactive progesterone receptor 23 kD UniProtKB:Q15185}; cPGES; cytosolic prostaglandin E synthase; Cytosolic prostaglandin E2 synthase; Hsp90 co-chaperone; p23 cochaperone; Progesterone receptor complex p23; Prostaglandin E synthase 3; prostaglandin E synthase 3 (cytosolic); prostaglandin-E synthase 3; ptges3 {ECO:0000250|UniProtKB:Q15185}; Sid 3177; telomerase binding protein, p23; Telomerase-binding protein p23; unactive progesterone receptor, 23 kD
Gene Aliases: 5730442A20Rik; cPGES; P23; Ptges; PTGES3; RGD1561913; Sid3177; TEBP
UniProt ID: (Human) Q15185, (Rat) P83868, (Mouse) Q9R0Q7
Entrez Gene ID: (Human) 10728, (Rat) 362809, (Mouse) 56351
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.