Invitrogen
Manufacturer Code:PA568800
Catalog # PIPA568800
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse APE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AP endonuclease 1; AP endonuclease 1 AP endonuclease class I AP lyase DNA-(apurinic or apyrimidinic site) lyase apurinic-apyrimidinic endonuclease 1 apurinic/apyrimidinic (abasic) endonuclease apurinic/apyrimidinic exonuclease deoxyribonuclease (apurinic or apyrimidinic) multifunctional DNA repair enzyme protein REF-1 redox factor 1; AP lyase; APEN; APEX nuclease; APEX nuclease (multifunctional DNA repair enzyme) 1; APEX nuclease 1; Apurinic-apyrimidinic endonuclease 1; apurinic/apyrimidinic endonuclease; apurinic/apyrimidinic endonuclease 1; BAP 1; DNA-(apurinic or apyrimidinic site) endonuclease; DNA-(apurinic or apyrimidinic site) endonuclease, mitochondrial; DNA-(apurinic or apyrimidinic site) lyase; Redox factor-1; REF-1
Gene Aliases: APE; APEN; APEX; APEX1; BAP1; HAP1; REF-1; REF1
UniProt ID: (Bovine) P23196, (Mouse) P28352, (Rat) P43138
Entrez Gene ID: (Rabbit) 100348924, (Horse) 100072588, (Bovine) 281630, (Mouse) 11792, (Guinea pig) 100719062, (Rat) 79116
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.