Invitrogen
Manufacturer Code:PA5116972
Catalog # PIPA5116972
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-318 of human APEX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AP endonuclease 1; AP endonuclease 1 AP endonuclease class I AP lyase APE APE1 APEN Apex APEX nuclease APEX nuclease (multifunctional DNA repair enzyme) 1 APEX nuclease 1 APEX1 apurinic/a Apurinic/apy apurinic/apyrimidinic (abasic) endonuclease apurinic/apyrimidinic endodeoxyribonuclease 1 apurinic/apyrimidinic endonuclease 1 Apurinic-apyrimidinic endonuclease 1 APX deoxyribonuclease (apurinic or apyrimidinic) DNA-(apurinic or apyrimidinic site) lyase DNA-(apurinic or apyrimidinic site) lyase mitochondrial HAP1 LOC100348924 protein REF-1 redox factor 1 redox factor-1 REF1 REF-1; AP endonuclease class I; AP lyase; APEN; APEX nuclease; APEX nuclease (multifunctional DNA repair enzyme) 1; Apurinic-apyrimidinic endonuclease 1; apurinic/apyrimidinic (abasic) endonuclease; deoxyribonuclease (apurinic or apyrimidinic); DNA-(apurinic or apyrimidinic site) endonuclease; DNA-(apurinic or apyrimidinic site) endonuclease, mitochondrial; protein REF-1; Redox factor-1; REF-1
Gene Aliases: APE; APE1; APEN; APEX; APEX1; APX; HAP1; REF1
UniProt ID: (Human) P27695
Entrez Gene ID: (Human) 328
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.