Microbiologics Helix Elite QC Panels and Molecular Standards
Microbiologics™ Helix Elite™ Quality Control Panels are designed to simulate patient samples in order to monitor the extraction, amplification, and detection process of molecular testing assays.
The panels offer a range of formats, from synthetic nucleic acid RNA and DNA sequences to inactivated, fully intact whole organisms. Targets covered include respiratory, women's health, STI, gastrointestinal, COVID, blood culture ID, and more.