Microbiologics EZ-Accu Shot Preparations
Microbiologics™ EZ-Accu™ Shot provides a fast, convenient, and reliable quantitative microorganism preparation for growth promotion testing, media challenge testing, and more.
Each instant-dissolve microorganism pellet is designed to deliver 10 to 100 CFU per inoculum and provide eight hours of stability after rehydration to help you meet U.S. Pharmacopeia guidelines.
NOTE: Please sign in to your fishersci.com account to view all of the products available for purchase.
Register for a fishersci.com account.