Learn More
Zymo Research Corporation Zyppy-96™ Plasmid Kit (8 x 96 Preps)
Supplier: Zymo Research Corporation D4043
The Zyppy-96 Plasmid Miniprep allows efficient isolation of plasmid DNA from E. coli, featuring a modified alkaline lysis system that is pellet-free. This completely bypasses tedious centrifugation, pelleting, and re-suspension steps common to conventional procedures. Instead, the uniquely formulated Deep Blue Lysis Buffer is added directly to the bacterial cultures in a 96-well block. Buffer neutralization and lysate separation steps are assisted with a specially designed Neutralization/Clearing Buffer. Eluted plasmid DNA is high quality, endotoxin-free, and well-suited for use in restriction endonuclease digestion, DNA ligation, PCR, transcription, sequencing, and other sensitive downstream applications including transfection.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.