Learn More
Zymo Research Corporation Zymoclean™ Gel DNA Recovery Kit (10 Preps) w/ Zymo-Spin™ I Columns (Uncapped)
SDP
Supplier:  Zymo Research Corporation D4001T
Encompass_Preferred
Simply add the specially formulated Agarose Dissolving Buffer (ADB) to the gel slice containing your DNA sample, let dissolve, and then transfer to the supplied Zymo-Spin™ Column. There is no need for organic denaturants or chloroform. Instead, the product utilizes Fast-Spin column technology to yield high-quality DNA in just 15 minutes. DNA purified using the Zymoclean™ Gel DNA Recovery Kit is perfectly suited for use in DNA ligation reactions, sequencing, DNA labeling reactions, PCR, etc.
Catalog No. 50-125-1374