Learn More
Zymo Research Corporation ZymoBroth™ (500 ml) ^
Supplier: Zymo Research Corporation M3015500
ZymoBroth (ZB) is a specially formulated growth medium used for the preparation of highly competent E. coli cells for DNA transformation.