Learn More
Zymo Research Corporation Zymo Environ Water RNA Kit (50 preps)
SDP
Supplier:  Zymo Research Corporation R2042
Encompass_Preferred
The use of Water Concentrating Buffer (WCB) promotes the pelleting of viral capsids and protein-associated RNA, increasing SARS-CoV-2 RNA yields by more than 8-fold. DNA/RNA Shield™ allows for safe handling as it has been validated for inactivation of pathogens including SARS-CoV-2. Finally, the workflow yields highly concentrated, PCR inhibitor-free RNA ready for downstream applications including RT-qPCR, RT-ddPCR, and NGS.
Catalog No. 50-203-7290
Explore available promotions