Learn More
ZOLL™ Medical AED Plus™ Fully Automatic Defibrillator Packages
Provides real CPR help for rate and depth of chest compressions
Supplier:  ZOLL™ Medical 800000401101
 View more versions of this product
Catalog No. 17-300-105
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
The AED Plus™ is the first and only full-rescue public access AED that provides Real CPR Help™ for rate and depth of chest compressions. Real-time feedback helps rescuers provide the best manual CPR possible in a sudden cardiac arrest (SCA). The AED Plus also guides rescuers through the complete Chain of Survival, helping all SCA victims, not just those who need a shock. The AED Plus features ZOLL's unique one-piece electrode pad for fast and accurate placement and is powered by lithium camera batteries available from retail stores. Both the pad and the batteries have a five-year shelf life.
  • Comes with Medical Prescription or PlusTrac Professional1 or PlusTrac Professional5
AED Plus w/Medical Prescription (Model 8000-004007-01) includes:
  • ZOLL Fully Automatic AED Plus w/AED Cover, PlusRX Medical Prescription, CPR-D padz™ Electrode, pack of 10 CR123a batteries, and Carry Case
  • AED Plus w/PlusTrac Professional1 (Model 8000-004010-01) includes:
  • ZOLL Fully Automatic AED Plus w/AED Cover, PlusTrac Professional1, CPR-D-padz Electrode, pack of 10 CR123a batteries, and Carry Case
  • AED Plus w/PlusTrac Professional5 (Model 8000-004011-01) includes:
  • ZOLL Fully Automatic AED Plus w/AED Cover, PlusTrac Professional5, CPR-D-padz Electrode, pack of 10 CR123a batteries, and Carry Case
Specifications
Specifications
Defibrillator Package
ZOLL fully automatic AED Plus w/AED cover, PlusTrac Professional5, CPR-D-padz electrode, pack of 10 batteries, and carry case