Learn More
Cole-Parmer YSI 3682 Zobell Solution for ORP Calibration, 1.25 g
Supplier: Cole-Parmer 0547860
Use Zobell solution by adding distilled water to verify the performance of your ORP electrodes.