Learn More
FIREBIRD OPTICS Zinc Selenide (ZnSe) Circular Optical Window- 25x4mm
Supplier: FIREBIRD OPTICS ZNSE25X4
Zinc Selenide (ZnSe) Circular Optical Window- 25x4mm• Wavelength range: 1-14µm •Surface quality: 60/40 • Clear aperture: 90% •Diameter tolerance: ±0.13mm •Thickness tolerance: ±0.13mm