Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coast Zithion-X AAA Rechargeable Batteries
Pack power with purpose. ZITHION-X™ rechargeable lithium batteries change the game in responsible, portable power technology.
$31.50 - $81.20
Specifications
Product Type | Rechargeable Battery |
---|---|
Battery Type | AAA |
Material | Lithium-Ion |
Cell Type | Spring Terminal |
Voltage | 1.5 V |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
17-100-474
|
Coast
30987 |
Each for $81.20
|
|
||||||
17-100-475
|
Coast
30986 |
Each for $59.60
|
|
||||||
17-100-476
|
Coast
30985 |
Each for $31.50
|
|
||||||
Description
- COAST ZITHION-X™ rechargeable lithium batteries change the game in responsible, portable power technology.
- Advanced engineering provides a comprehensive upgrade compared with other sources of battery power by delivering long-lasting run times for a clear performance advantage over standard rechargeable lithium and NiMH batteries.
- To mimic the broad compatibility of alkaline batteries, proprietary COAST ZX Optimized Voltage™ allows ZITHION-X batteries to modulate their power supply up or down based on the need and prevent flickering, overheating, and damage to products.
- The leak-resistant battery casing prevents corrosion during use, matching performance with protection.
- Connect conveniently via USB-C to recharge quickly (in 2.5 hours or less for AA and 1.25 hours or less for AAA) and forget about special charging cables and cradles. Every ZITHION-X can be recharged at least 1000 times, resulting in significant cost-savings compared with single-use disposable batteries while keeping hundreds of batteries out of the landfill.
Specifications
Rechargeable Battery | |
Lithium-Ion | |
1.5 V |
AAA | |
Spring Terminal |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title