Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Zippy Dissecting Kit w/ Screw-Lock with Blade Scalpel:(8 PCS)
SDP

The best dissecting kits for your needs!

Catalog No. 13-820-091



Add to Cart
This item is not returnable. View return policy

Description

Description

Fisherbrand™ Zippy Dissecting Kit w/ Screw-Lock with Blade Scalpel:(8 PCS)

  • Red zippered case-6 Loop;
  • 6 in. student Dissecting Scalpel: Screw-lock w/ Blade
  • 4-1/2 in. Student Dissecting Scissors- Sharp Point
  • 4-1/2 in. Student Forceps:
  • Blunt, Serr. Tips; Serr. Grip
  • Straight teasing Needle w/ 4 in.Plastic Handle
  • Angular teasing Needle w/ 4 in. Plastic Handle
  • Glass dropper w/ rubber bulb- large Size 5 in.
  • 6 in. hard Plastic ruler

Certifications

ISO 9001-02, GMP, cGMP

Specifications

Specifications

8 Pcs
Dissecting Experiments
8 Pc.
Dissection tool Kit
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit