Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc Metal, 6 x 1 x 0.019 in., Strips, 99.8%, Spectrum™ Chemical

Click to view available options
Quantity:
500 g
Packaging:
Amber Glass Bottle
Specifications
CAS Max % | 1 |
Quantity | 500 g |
Percent Purity | 99.8% |
Grade | Reagent |
Packaging | Amber Glass Bottle |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.