Learn More
Epredia™ Zinc Formalin Specimen Transportation System - STS™ Prefilled Specimen Containers
Transport, store and retrieve pathology specimens in a safe and cost effective manner with the Epredia™ Zinc Formalin Specimen Transportation System - STS™ Prefilled Specimen Containers.
Supplier: Epredia™ 599601ZF9
Description
Closure System- Patented Bio-Tite™ closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
- Nonsterile containers marked with a lot code and expiration date
- Comply with all updated federal and state OSHA requirement
- Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
Specifications
Polypropylene | |
960 mL, pre-filled 480 mL | |
Clear | |
9/Cs. |
Non-sterile | |
Snap Cap | |
Prefilled containers with 10% Zinc Formalin |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.