Learn More
Polysciences, Inc. Zinc Formalin Fixative, pH 6.25, 3.75 L
SDP
Supplier:  Polysciences, Inc. 215163.75L
Encompass
Excellent morphological preservation of nuclear & cytoplasmic components. Can replace neutral buffered formalin for routine tissue and immunohistochemical procedures. Non-precipitating fixative which can be used with automated and manual methods. Zinc formalin is best used as a primary fixative which minimizes denaturation. Specimens initially fixed in buffered formalin may be post fixed in Zinc Formalin for a week without an effect.
Catalog No. NC1744632
Explore available promotions