Learn More
Perkin Elmer US LLC Zero Dilution Glass Outer Liner for PSS Injectors
Supplier:  Perkin Elmer US LLC N1011447
Encompass_Preferred
Inlet liners for split injection have mixing chambers with tortuous flow paths to allow full vaporization of the sample. Deactivating the surface of these liners prevents active compounds from degrading. Packing the liner with wool will trap non-volatile residue and prevent column contamination when analyzing dirty samples. Not compatible with the Clarus 590/690 Capillary Injectors.
Catalog No. 50-210-4144