Learn More
Mti Corporation ZERO DIFFRACTION PLATE FOR XRD
Supplier: Mti Corporation ZEROSI24D10C1CAVITY15D
Zero Diffraction Plate for XRD sample: 24.6 Dia x 1.0 t mm with Cavity 15 ID x 0.5 mm, Si Crystal