Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix NATtrol EBV External Run Control Strain: B95-8
Designed to evaluate the performance of nucleic acid tests for determination of the presence of EBV DNA
$433.00 - $523.00
Specifications
For Use With (Application) | Designed to evaluate the performance of nucleic acid tests for determination of the presence of EBV DNA, can also be used for validation of clinical assays, development of diagnostic tests and training of laboratory personnel |
---|---|
Specificity | Evaluate the performance of Nucleic Acid |
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
21-020-110
|
ZeptoMetrix
NATEBVERCL |
External Run Control (low) |
Each for $433.00
|
|
|||||
21-020-111
|
ZeptoMetrix
NATEBVERCM |
External Run Control (medium) |
Each for $523.00
|
|
|||||
Description
- Formulated with purified, intact virus particles that have been chemically modified to render them non-infectious and refrigerator stable
- Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
Specifications
Designed to evaluate the performance of nucleic acid tests for determination of the presence of EBV DNA, can also be used for validation of clinical assays, development of diagnostic tests and training of laboratory personnel | |
Evaluate the performance of Nucleic Acid |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title